Assay Details
Target Gene Details
Entrez Gene ID: | 13875 |
Gene Name: | Ets2 repressor factor |
Gene Aliases: |
- |
Location: |
Chr.7:25242560-25250758 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 3 - Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Erf | NM_010155.3 | NP_034285.3 | ||
| AK159225.1 | BAE34912.1 | |||
| BC053045.1 | AAH53045.1 | |||
| BC059176.1 | AAH59176.1 | |||
| BC063092.1 | AAH63092.1 |
More Information
Set Membership: |
|

Genomic Map