Assay Details
Target Gene Details
Entrez Gene ID: | 15464 |
Gene Name: | histidine rich calcium binding protein |
Gene Aliases: |
- |
Location: |
Chr.7:45335269-45338973 on Build GRCm38 |
Assay Gene Location: | Within Intron 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Hrc | NM_010473.2 | NP_034603.2 | ||
| AF132218.1 | AAD42061.1 | |||
| AF158597.1 | AAD55250.1 | |||
| AI322629.1 | ||||
| BC141121.1 |
More Information
Set Membership: |
|

Genomic Map