Assay Details
Target Gene Details
Entrez Gene ID: | 110033 |
Gene Name: | kinesin family member 22 |
Gene Aliases: |
AU021460, C81217, Kid, Kif22a |
Location: |
Chr.7:127027729-127042466 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 12 - Exon 12 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Kif22 | NM_145588.1 | NP_663563.1 | ||
| AK075922.1 | BAE43356.1 | |||
| AK084310.1 | ||||
| AK088033.1 | ||||
| AK205411.1 | ||||
| BC003427.1 | AAH03427.1 |
More Information
Set Membership: |
|

Genomic Map