Assay Details
Target Gene Details
Entrez Gene ID: | 381853 |
Gene Name: | gastric inhibitory polypeptide receptor |
Gene Aliases: |
GIP-R, Gm1081, Gm160 |
Location: |
Chr.7:19156063-19170248 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 7 - Exon 8 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Gipr | NM_001080815.1 | NP_001074284.1 | ||
| AK133386.1 | ||||
| AK146552.1 |
More Information
Set Membership: |
|

Genomic Map