Assay Details
Target Gene Details
Entrez Gene ID: | 232801 |
Gene Name: | leukocyte immunoglobulin-like receptor, subfamily A (with TM domain), member 5 |
Gene Aliases: |
E030017K20, EG232801, Gm4878, Lilrc1 |
Location: |
Chr.7:4237643-4243690 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 1 - Intron 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Lilra5 | NM_001081239.2 | NP_001074708.2 | ||
| AY998482.1 | AAY44811.1 |
More Information
Set Membership: |
|

Genomic Map