Assay Details
Target Gene Details
Entrez Gene ID: | 243912 |
Gene Name: | heat shock protein, alpha-crystallin-related, B6 |
Gene Aliases: |
AA387366, Gm479, Hsp20 |
Location: |
Chr.7:30552634-30555439 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 1 - Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Hspb6 | NM_001012401.2 | NP_001012401.1 | ||
| AK144390.1 | BAE25863.1 | |||
| BC089621.1 | AAH89621.1 |

Genomic Map