Assay Details
Target Gene Details
Entrez Gene ID: | 13593 |
Gene Name: | early B cell factor 3 |
Gene Aliases: |
3110018A08Rik, O/E-2, mKIAA4201 |
Location: |
Chr.7:137193671-137322810 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 6 - Exon 7 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Ebf3 | NM_001113414.1 | NP_001106885.1 | ||
| NM_001113415.1 | NP_001106886.1 | |||
| NM_010096.3 | NP_034226.1 | |||
| AK138602.1 | BAE23711.1 | |||
| AK220542.1 | BAD90316.1 | |||
| BC067018.1 | AAH67018.1 | |||
| U82442.1 | AAC64323.1 | |||
| U92702.1 | AAB58322.1 | |||
| U92704.1 | AAB58324.1 |
Target Gene Details
Entrez Gene ID: | 100038555 |
Gene Name: | predicted gene 10578 |
Gene Aliases: |
ENSMUSG00000073800 |
Location: |
Chr.7:137234683-137242609 on Build GRCm38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Gm10578 | NR_045885.1 | |||
| AK133282.1 | BAE21591.1 |
More Information
Set Membership: |
|

Genomic Map