Assay Details
Target Gene Details
Entrez Gene ID: | 56188 |
Gene Name: | FXYD domain-containing ion transport regulator 1 |
Gene Aliases: |
0610012C17Rik, 1110006M24Rik, Plm, Pml |
Location: |
Chr.7:31051678-31055877 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 2 - Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Fxyd1 | NM_019503.4 | NP_062376.2 | ||
| NM_052991.4 | NP_443717.1 | |||
| NM_052992.3 | NP_443718.1 | |||
| NM_194321.2 | NP_919302.1 | |||
| AA462069.1 | ||||
| AA466595.1 | ||||
| AF089734.1 | AAD41683.1 | |||
| AK002585.1 | BAB22208.1 | |||
| BC024671.1 | AAH24671.1 | |||
| BY064152.1 | ||||
| BY132051.1 | ||||
| CA459152.1 | ||||
| CK793316.1 | ||||
| W16239.1 | ||||
| W98524.1 |
More Information
Set Membership: |
|

Genomic Map