Assay Details
Target Gene Details
Entrez Gene ID: | 68077 |
Gene Name: | glioma tumor suppressor candidate region gene 2 |
Gene Aliases: |
5330430H08Rik, 9430097C02Rik, AU041936, AW536441, PICT-1, R74911 |
Location: |
Chr.7:15937835-15946108 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 6 - Exon 7 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Gltscr2 | NM_133831.3 | NP_598592.2 | ||
| AK077341.1 | BAC36760.1 | |||
| AK079167.1 | ||||
| AK088461.1 | BAC40367.1 | |||
| AK146916.1 | BAE27528.1 | |||
| AK151836.1 | BAE30729.1 | |||
| AK158665.1 | ||||
| AK160070.1 | BAE35605.1 | |||
| AK169290.1 | BAE41047.1 | |||
| AY535001.1 | AAS46029.1 | |||
| BC017637.1 | AAH17637.1 | |||
| BC025810.1 | AAH25810.1 |

Genomic Map