Assay Details
Target Gene Details
Entrez Gene ID: | 101612 |
Gene Name: | glutamate-rich WD repeat containing 1 |
Gene Aliases: |
A301, AI504353, Grwd, Wdr28 |
Location: |
Chr.7:45825223-45830796 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 1 - Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Grwd1 | NM_153419.2 | NP_700468.2 | ||
| AB001539.1 | BAC66461.1 | |||
| AK077611.1 | BAC36899.1 | |||
| BC008121.1 | AAH08121.1 | |||
| BC028896.1 | AAH28896.1 | |||
| BC083143.1 | AAH83143.1 | |||
| BQ886252.1 |

Genomic Map