Assay Details
Target Gene Details
Entrez Gene ID: | 243914 |
Gene Name: | leucine-rich repeat LGI family, member 4 |
Gene Aliases: |
Lgil3, clp |
Location: |
Chr.7:31058953-31070935 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 2 - Intron 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Lgi4 | NM_144556.2 | NP_653139.2 | ||
| AJ487521.1 | CAD31789.1 | |||
| AJ487960.1 | CAD32307.1 | |||
| AK033967.1 | ||||
| AK134643.1 | BAE22223.1 | |||
| AK148793.1 | ||||
| BC098469.1 | AAH98469.1 |
More Information
Set Membership: |
|

Genomic Map