Assay Details
Target Gene Details
Entrez Gene ID: | 56312 |
Gene Name: | nuclear protein transcription regulator 1 |
Gene Aliases: |
2310032H04Rik, Com1, p8 |
Location: |
Chr.7:126623246-126625470 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 2 - Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Nupr1 | NM_019738.1 | NP_062712.1 | ||
| AF131196.1 | AAD41785.1 | |||
| AK009572.1 | BAB26369.1 | |||
| BC002109.1 | AAH02109.1 |

Genomic Map