Assay Details
Target Gene Details
Entrez Gene ID: | 57296 |
Gene Name: | proteasome (prosome, macropain) 26S subunit, non-ATPase, 8 |
Gene Aliases: |
6720456J22Rik, AA407360, AL033291, AL033322, AL033323, C76433 |
Location: |
Chr.7:29174187-29180673 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 5 - Exon 6 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Psmd8 | NM_026545.3 | NP_080821.3 | ||
| AK002921.1 | BAB22458.2 | |||
| AK003436.1 | BAB22789.2 | |||
| AK020132.1 | BAB32006.2 | |||
| AK027973.1 | BAC25683.1 | |||
| AK051257.1 | BAC34576.1 | |||
| AK150493.1 | BAE29609.1 | |||
| AK150982.1 | BAE30009.1 | |||
| AK153329.1 | BAE31908.1 | |||
| AK159216.1 | BAE34905.1 | |||
| AK159795.1 | BAE35376.1 | |||
| AK159926.1 | BAE35488.1 | |||
| AK166718.1 | BAE38968.1 | |||
| AK167948.1 | BAE39951.1 | |||
| AK168886.1 | BAE40703.1 | |||
| BC004075.1 | AAH04075.1 | |||
| BC005717.1 | AAH05717.1 | |||
| CD774979.1 |
More Information
Set Membership: |
|

Genomic Map