Assay Details
Target Gene Details
Entrez Gene ID: | 18733 |
Gene Name: | paired Ig-like receptor B |
Gene Aliases: |
Gp91, LIR-3, Lilrb3, PIR-B |
Location: |
Chr.7:3712505-3720456 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 9 - Exon 9 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Pirb | NM_011095.2 | NP_035225.2 | ||
| AF148880.1 | AAD41829.1 | |||
| AK142375.1 | BAE25046.1 | |||
| AK152232.1 | ||||
| AK153116.1 | ||||
| AK165255.1 | BAE38107.1 | |||
| AK170618.1 | ||||
| AK170720.1 | ||||
| U83172.1 | AAB40934.1 | |||
| U96689.1 | AAC53219.1 | |||
| U96692.1 | AAC53222.1 | |||
| U96693.1 | AAC53223.1 |
More Information
Set Membership: |
|

Genomic Map