Assay Details
Target Gene Details
Entrez Gene ID: | 243912 |
Gene Name: | heat shock protein, alpha-crystallin-related, B6 |
Gene Aliases: |
AA387366, Gm479, Hsp20 |
Location: |
Chr.7:30552634-30555439 on Build GRCm38 |
Assay Gene Location: | Overlaps - Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Hspb6 | NM_001012401.2 | NP_001012401.1 | ||
| AK144390.1 | BAE25863.1 | |||
| BC089621.1 | AAH89621.1 |
Target Gene Details
Entrez Gene ID: | 75660 |
Gene Name: | lin-37 homolog (C. elegans) |
Gene Aliases: |
1810054G18Rik |
Location: |
Chr.7:30555441-30559719 on Build GRCm38 |
Assay Gene Location: | Overlaps - Exon 9 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Lin37 | NM_001290569.1 | NP_001277498.1 | ||
| NM_029377.2 | NP_083653.1 | |||
| AK007858.1 | BAB25310.1 | |||
| BC013546.1 | AAH13546.1 |

Genomic Map