Assay Details
Target Gene Details
Entrez Gene ID: | 17907 |
Gene Name: | myosin light chain, phosphorylatable, fast skeletal muscle |
Gene Aliases: |
2410014J02Rik, MLC-2, Mlc2 |
Location: |
Chr.7:127208848-127214288 on Build GRCm38 |
Assay Gene Location: | Overlaps - Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Mylpf | NM_016754.5 | NP_058034.1 | ||
| AK010483.1 | BAB26974.1 |
More Information
Set Membership: |
|

Genomic Map