Assay Details
Target Gene Details
Entrez Gene ID: | 75212 |
Gene Name: | ring finger protein 121 |
Gene Aliases: |
4930544L10Rik |
Location: |
Chr.7:102019145-102072428 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 8 - Exon 8 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Rnf121 | NM_029211.2 | NP_083487.2 | ||
| AK016037.1 | BAB30089.2 | |||
| AK034806.1 | BAC28837.1 | |||
| AK078509.1 | BAC37314.1 | |||
| AK159032.1 | BAE34777.1 | |||
| AK159260.1 | BAE34941.1 | |||
| AK168821.1 | BAE40648.1 | |||
| AK169089.1 | BAE40873.1 | |||
| BC022686.1 | AAH22686.1 |
More Information
Set Membership: |
|

Genomic Map