Assay Details
Target Gene Details
Entrez Gene ID: | 244238 |
Gene Name: | MAS-related GPR, member E |
Gene Aliases: |
C130069N09Rik, Ebrt3, Mgrf, MrgE |
Location: |
Chr.7:143778363-143784500 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 1 - Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Mrgpre | NM_175534.3 | NP_780743.1 | ||
| AJ320231.1 | CAC86257.1 | |||
| AK048528.1 | BAC33361.1 | |||
| AK133657.1 | BAE21769.1 | |||
| AK154886.1 | BAE32903.1 | |||
| BC096766.1 | AAH96766.1 | |||
| BC138305.1 | ||||
| BC138308.1 |
More Information
Set Membership: |
|

Genomic Map