Assay Details
Target Gene Details
Entrez Gene ID: | 381974 |
Gene Name: | MAS-related GPR, member G |
Gene Aliases: |
Gm1098, Mrgg |
Location: |
Chr.7:143763710-143766993 on Build GRCm38 |
Assay Gene Location: | Overlaps - Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Mrgprg | NM_203492.2 | NP_987077.1 | ||
| BC100302.1 | AAI00303.1 |
More Information
Set Membership: |
|

Genomic Map