Assay Details
Target Gene Details
Entrez Gene ID: | 102466219 |
Gene Name: | microRNA 7061 |
Gene Aliases: |
mmu-mir-7061 |
Location: |
Chr.7:130908268-130908335 on Build GRCm38 |
Assay Gene Location: | Overlaps - Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Mir7061 | NR_106028.1 |
Target Gene Details
Entrez Gene ID: | 101476 |
Gene Name: | pleckstrin homology domain containing, family A (phosphoinositide binding specific) member 1 |
Gene Aliases: |
AA960558, C920009D07Rik, TAPP1 |
Location: |
Chr.7:130865729-130913308 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 8 - Exon 9 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Plekha1 | NM_133942.2 | NP_598703.1 | ||
| AK044224.1 | ||||
| AK083340.1 | BAC38874.1 | |||
| BC020017.1 | AAH20017.1 |

Genomic Map