Assay Details
Target Gene Details
Entrez Gene ID: | 381925 |
Gene Name: | phosphatidic acid phosphatase type 2 domain containing 1A |
Gene Aliases: |
C030048B12Rik, Gm1090 |
Location: |
Chr.7:129257094-129391307 on Build GRCm38 |
Assay Gene Location: | Overlaps Exon 1 - Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Ppapdc1a | NM_001080963.1 | NP_001074432.1 | ||
| AK081292.1 | ||||
| BC120497.1 |
More Information
Set Membership: |
|

Genomic Map