Assay Details
Target Gene Details
Entrez Gene ID: | 15469 |
Gene Name: | protein arginine N-methyltransferase 1 |
Gene Aliases: |
6720434D09Rik, AW214366, Hrmt1l2, Mrmt1 |
Location: |
Chr.7:44976755-44986420 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 11 - Exon 11 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Prmt1 | NM_001252476.1 | NP_001239405.1 | ||
| NM_001252477.1 | NP_001239406.1 | |||
| NM_019830.3 | NP_062804.1 | |||
| NR_045521.1 | ||||
| AF232716.1 | AAF37292.1 | |||
| AF232717.1 | AAF37293.1 | |||
| AI846916.1 | ||||
| AK020120.1 | BAB32002.1 | |||
| AK088790.1 | BAC40573.1 | |||
| AK146975.1 | ||||
| AK152342.1 | ||||
| AK164421.1 | BAE37781.1 | |||
| AK182201.1 | ||||
| BC002249.1 | AAH02249.1 | |||
| BC051547.1 | AAH51547.1 | |||
| BC051953.1 | AAH51953.1 | |||
| BC062964.1 | AAH62964.1 | |||
| BY760714.1 |
More Information
Set Membership: |
|

Genomic Map