Assay Details
Target Gene Details
Entrez Gene ID: | 67308 |
Gene Name: | mitochondrial ribosomal protein L46 |
Gene Aliases: |
3110052F15Rik, C15orf4, C80107, LIECG2, P2ECSL |
Location: |
Chr.7:78775341-78783089 on Build GRCm38 |
Assay Gene Location: | Within Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Mrpl46 | NM_023331.2 | 2 | 340 | NP_075820.1 |
| AF217090.1 | 2 | 307 | AAG44099.1 | |
| AK006415.1 | BAB24577.1 | |||
| AK014205.1 | 2 | 317 | ||
| BC061094.1 | 2 | 340 | AAH61094.1 |
More Information
Set Membership: |
|

Genomic Map