Assay Details
Target Gene Details
Entrez Gene ID: | 258961 |
Gene Name: | olfactory receptor 631 |
Gene Aliases: |
MOR3-1 |
Location: |
Chr.7:103904306-103930666 on Build GRCm38 |
Assay Gene Location: | Overlaps Exon 2 - Intron 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Olfr631 | NM_001271020.1 | NP_001257949.1 | ||
| NM_146959.2 | NP_667170.2 |
More Information
Set Membership: |
|

Genomic Map