Assay Details
Target Gene Details
Entrez Gene ID: | 66786 |
Gene Name: | olfactory receptor 701 |
Gene Aliases: |
4932441H21Rik, 4933433E02Rik, MOR283-1 |
Location: |
Chr.7:106814144-106820709 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 2 - Intron 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Olfr701 | NM_028910.3 | NP_083186.2 | ||
| AK015248.1 | BAB29764.1 | |||
| AK016560.1 | BAB30304.1 | |||
| AK017036.1 | BAB30564.1 | |||
| BC110461.2 | AAI10462.1 |
More Information
Set Membership: |
|

Genomic Map