Assay Details
Target Gene Details
Entrez Gene ID: | 170639 |
Gene Name: | olfactory receptor 78 |
Gene Aliases: |
4633402A21Rik, MOL2.3, MOR18-2, Or51e2, PSGR, RA1c |
Location: |
Chr.7:102740721-102759471 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 1 - Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Olfr78 | NM_001168503.1 | NP_001161975.1 | ||
| NM_130866.4 | NP_570936.1 | |||
| AK014616.1 | ||||
| AK028467.1 | BAC25966.1 | |||
| AK036356.1 | BAC29396.1 |
More Information
Set Membership: |
|

Genomic Map