Assay Details
Target Gene Details
Entrez Gene ID: | 243910 |
Gene Name: | nuclear factor of kappa light polypeptide gene enhancer in B cells inhibitor, delta |
Gene Aliases: |
IkappaBNS |
Location: |
Chr.7:30421729-30428746 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 5 - Exon 6 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Nfkbid | NM_172142.3 | NP_742154.1 | ||
| AK155969.1 | ||||
| AK167356.1 | BAE39455.1 | |||
| AY078069.1 | AAL79957.1 | |||
| BC110633.1 | AAI10634.1 |
More Information
Set Membership: |
|

Genomic Map