Assay Details
Target Gene Details
Entrez Gene ID: | 107702 |
Gene Name: | ribonuclease/angiogenin inhibitor 1 |
Gene Aliases: |
AW546468, C80305, RNH |
Location: |
Chr.7:141160326-141172851 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 5 - Exon 6 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Rnh1 | NM_001172100.1 | NP_001165571.1 | ||
| NM_001172101.1 | NP_001165572.1 | |||
| NM_145135.3 | NP_660117.2 | |||
| AF071546.1 | AAK68859.1 | |||
| AK133822.1 | BAE21862.1 | |||
| AK141351.1 | BAE24659.1 | |||
| AK145173.1 | BAE26275.1 | |||
| AK147059.1 | BAE27643.1 | |||
| BC010331.1 | AAH10331.1 |

Genomic Map