Assay Details
Target Gene Details
Entrez Gene ID: | 19342 |
Gene Name: | RAB4B, member RAS oncogene family |
Gene Aliases: |
1500031G17Rik |
Location: |
Chr.7:27168424-27178901 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 7 - Exon 7 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Rab4b | NM_029391.2 | NP_083667.1 | ||
| AB232592.1 | BAF02854.1 | |||
| AF408432.1 | AAL11725.1 | |||
| AK005314.1 | BAB23948.1 | |||
| AK140513.1 | BAE24410.1 | |||
| AK170552.1 | BAE41875.1 | |||
| BC007147.1 | AAH07147.1 | |||
| BC096654.1 | AAH96654.1 |

Genomic Map