Assay Details
Target Gene Details
Entrez Gene ID: | 100504184 |
Gene Name: | predicted gene 16336 |
Gene Aliases: |
OTTMUSG00000033852 |
Location: |
Chr.7:110904985-110905851 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 1 - Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Gm16336 | NR_045267.1 | |||
| BE633821.1 |
Target Gene Details
Entrez Gene ID: | 17540 |
Gene Name: | MRV integration site 1 |
Gene Aliases: |
BB115629, Irag, Ris1 |
Location: |
Chr.7:110868257-110982908 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 11 - Exon 11 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Mrvi1 | NM_010826.5 | NP_034956.2 | ||
| NM_194464.3 | NP_919446.2 | |||
| AK141821.1 | BAE24844.1 | |||
| AK157169.1 | BAE33986.1 | |||
| U63407.1 | AAD22568.1 | |||
| U63408.1 | AAD22569.1 |
More Information
Set Membership: |
|

Genomic Map