Assay Details
Target Gene Details
Entrez Gene ID: | 233046 |
Gene Name: | RAS guanyl releasing protein 4 |
Gene Aliases: |
- |
Location: |
Chr.7:29134933-29153961 on Build GRCm38 |
Assay Gene Location: | Overlaps Exon 1 - Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Rasgrp4 | NM_001174155.1 | NP_001167626.1 | ||
| NM_145149.4 | NP_660131.2 | |||
| NR_045676.1 | ||||
| AF331457.1 | AAK84299.1 | |||
| AK087348.1 | ||||
| AK089282.1 | BAC40827.1 | |||
| AK154746.1 | BAE32802.1 | |||
| AK162667.1 | BAE37013.1 | |||
| AY040628.1 | AAK81694.1 | |||
| AY196476.1 | AAO20841.1 | |||
| BY749980.1 |
More Information
Set Membership: |
|

Genomic Map