Assay Details
Target Gene Details
Entrez Gene ID: | 212508 |
Gene Name: | mitochondrial GTPase 1 homolog (S. cerevisiae) |
Gene Aliases: |
Gm169, Gtpbp7 |
Location: |
Chr.7:140137564-140150786 on Build GRCm38 |
Assay Gene Location: | Overlaps Exon 4 - Intron 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Mtg1 | NM_199301.2 | NP_955005.2 | ||
| AK144453.1 | BAE25897.1 | |||
| AK144625.1 | BAE25976.1 | |||
| BC027306.1 | AAH27306.1 | |||
| BY240360.1 |

Genomic Map