Assay Details
Target Gene Details
Entrez Gene ID: | 80749 |
Gene Name: | leucine rich repeat and fibronectin type III domain containing 1 |
Gene Aliases: |
BC004018, SALM2, Semo1 |
Location: |
Chr.7:28451985-28468242 on Build GRCm38 |
Assay Gene Location: | Overlaps Exon 3 - Intron 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Lrfn1 | NM_001141921.1 | NP_001135393.1 | ||
| NM_030562.2 | NP_085039.2 | |||
| AK090175.1 | BAC41123.1 | |||
| AK144493.1 | ||||
| BC004018.1 | AAH04018.1 |

Genomic Map