Assay Details
Target Gene Details
Entrez Gene ID: | 17136 |
Gene Name: | myelin-associated glycoprotein |
Gene Aliases: |
Gma, siglec-4a |
Location: |
Chr.7:30899175-30914922 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 10 - Intron 11 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Mag | NM_010758.2 | NP_034888.1 | ||
| BC101944.1 | AAI01945.1 | |||
| BC103521.1 | AAI03522.1 | |||
| BC103522.1 | AAI03523.1 | |||
| BC103523.1 | AAI03524.1 | |||
| M31811.1 | AAA39487.1 |
More Information
Set Membership: |
|

Genomic Map