Assay Details
Target Gene Details
Entrez Gene ID: | 52377 |
Gene Name: | reticulocalbin 3, EF-hand calcium binding domain |
Gene Aliases: |
6030455P07Rik, D530026G20Rik, D7Ertd671e, RLP49 |
Location: |
Chr.7:45082914-45092257 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Rcn3 | NM_026555.2 | 1 | 60 | NP_080831.2 |
| AK077943.1 | BAC37077.1 | |||
| AK082762.1 | BAC38608.1 | |||
| AK086705.1 | ||||
| AK132143.1 | ||||
| BC025602.1 | AAH25602.1 | |||
| BC055903.1 | AAH55903.1 | |||
| BY248337.1 | 1 | 62 |
More Information
Set Membership: |
|

Genomic Map