Assay Details
Target Gene Details
Entrez Gene ID: | 107503 |
Gene Name: | activating transcription factor 5 |
Gene Aliases: |
AFTA, Atf7, Atfx, ODA-10 |
Location: |
Chr.7:44812256-44816658 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 1 - Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Atf5 | NR_033136.1 | |||
| AF310624.1 | AAL55820.1 | |||
| AF312938.1 | AAG34160.1 | |||
| AF375475.1 | AAK54640.1 | |||
| AK131939.1 | ||||
| AK146479.1 | BAE27202.1 | |||
| AK159624.1 | ||||
| BC092068.1 | AAH92068.1 | |||
| BY307992.1 |
Target Gene Details
Entrez Gene ID: | 100328588 |
Gene Name: | Nup62-Il4i1 protein |
Gene Aliases: |
- |
Location: |
Chr.7:44816370-44840804 on Build GRCm38 |
Assay Gene Location: | Overlaps - Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Nup62-il4i1 | NM_001171024.1 | NP_001164495.1 | ||
| AK133082.1 | BAE21502.1 | |||
| BY087056.1 | ||||
| BY088421.1 | ||||
| CJ047381.1 |
More Information
Set Membership: |
|

Genomic Map