Assay Details
Target Gene Details
Entrez Gene ID: | 12587 |
Gene Name: | melanoma inhibitory activity |
Gene Aliases: |
CD-RAP, Cdrap, Mia1 |
Location: |
Chr.7:27179741-27181149 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Mia | NM_019394.3 | 1 | 75 | NP_062267.2 |
| AK135151.1 | BAE22443.1 | |||
| BE864464.1 | 1 | 75 | ||
| CK794645.1 | 1 | 69 | ||
| X94322.1 | 1 | 80 | CAA63983.1 |

Genomic Map