Assay Details
Target Gene Details
Entrez Gene ID: | 244237 |
Gene Name: | tumor necrosis factor receptor superfamily, member 26 |
Gene Aliases: |
Tnfrh3, Tnfrsf24 |
Location: |
Chr.7:143607685-143627845 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 6 - Exon 6 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Tnfrsf26 | NM_175649.5 | NP_783580.1 | ||
| AK154666.1 | BAE32753.1 | |||
| AY165628.1 | AAN87808.1 | |||
| BC111108.1 | AAI11109.1 | |||
| BC138819.1 | ||||
| BC138820.1 | ||||
| BC152357.1 | ||||
| BC152358.1 |
More Information
Set Membership: |
|

Genomic Map