Assay Details
Target Gene Details
Entrez Gene ID: | 21953 |
Gene Name: | troponin I, skeletal, fast 2 |
Gene Aliases: |
- |
Location: |
Chr.7:142440494-142444410 on Build GRCm38 |
Assay Gene Location: | Overlaps Exon 6 - Intron 6 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Tnni2 | NM_009405.2 | NP_033431.1 | ||
| BC028515.1 | AAH28515.1 | |||
| J04992.1 | AAA40485.1 |
More Information
Set Membership: |
|

Genomic Map