Assay Details
Target Gene Details
Entrez Gene ID: | 21953 |
Gene Name: | troponin I, skeletal, fast 2 |
Gene Aliases: |
- |
Location: |
Chr.7:142440494-142444410 on Build GRCm38 |
Assay Gene Location: | Within Exon 7 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Tnni2 | NM_009405.2 | 7 | 533 | NP_033431.1 |
| BC028515.1 | 8 | 571 | AAH28515.1 | |
| J04992.1 | 7 | 533 | AAA40485.1 |
More Information
Set Membership: |
|

Genomic Map