Assay Details
Target Gene Details
Entrez Gene ID: | 75212 |
Gene Name: | ring finger protein 121 |
Gene Aliases: |
4930544L10Rik |
Location: |
Chr.7:102019145-102072428 on Build GRCm38 |
Assay Gene Location: |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Rnf121 | AK016037.1 | BAB30089.2 |
Target Gene Details
Entrez Gene ID: | 102443350 |
Gene Name: | Xrcc1 N-terminal domain containing 1 |
Gene Aliases: |
Xndr |
Location: |
Chr.7:102065491-102083762 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Xndc1 | NM_001286689.1 | 1 | 222 | NP_001273618.1 |
| NM_001286690.1 | 1 | 222 | NP_001273619.1 | |
| BC067003.1 | 1 | 202 | AAH67003.1 |
Target Gene Details
Entrez Gene ID: | 102443351 |
Gene Name: | Xndc1-transient receptor potential cation channel, subfamily C, member 2 readthrough |
Gene Aliases: |
Xndr-trpc2 |
Location: |
Chr.7:102065491-102096864 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Xntrpc | NR_104582.1 | 1 | 222 | |
| AK040538.1 |

Genomic Map