Assay Details
Target Gene Details
Entrez Gene ID: | 102443350 |
Gene Name: | Xrcc1 N-terminal domain containing 1 |
Gene Aliases: |
Xndr |
Location: |
Chr.7:102065491-102083762 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 10 - Exon 11 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Xndc1 | NM_001286689.1 | NP_001273618.1 | ||
| NM_001286690.1 | NP_001273619.1 | |||
| AK041300.1 | BAC30894.1 | |||
| BC067003.1 | AAH67003.1 |
Target Gene Details
Entrez Gene ID: | 102443351 |
Gene Name: | Xndc1-transient receptor potential cation channel, subfamily C, member 2 readthrough |
Gene Aliases: |
Xndr-trpc2 |
Location: |
Chr.7:102065491-102096864 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 9 - Exon 10 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Xntrpc | NM_011644.3 | NP_035774.2 | ||
| NR_104582.1 | ||||
| AK040538.1 |

Genomic Map