Assay Details
Target Gene Details
Entrez Gene ID: | 21954 |
Gene Name: | troponin I, cardiac 3 |
Gene Aliases: |
Tn1, cTnI |
Location: |
Chr.7:4518308-4522443 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 6 - Exon 7 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Tnni3 | NM_009406.3 | NP_033432.1 | ||
| AK169223.1 | BAE40992.1 | |||
| BC061171.1 | AAH61171.1 | |||
| BC100590.1 | AAI00591.1 | |||
| U09181.1 | AAA19657.1 |
More Information
Set Membership: |
|

Genomic Map