Assay Details
Target Gene Details
Entrez Gene ID: | 21955 |
Gene Name: | troponin T1, skeletal, slow |
Gene Aliases: |
AW146156, Tnt, sTnT, ssTnT |
Location: |
Chr.7:4504570-4515999 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 2 - Intron 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Tnnt1 | NM_001277903.1 | NP_001264832.1 | ||
| NM_001277904.1 | NP_001264833.1 | |||
| NM_011618.2 | NP_035748.1 | |||
| AF020946.1 | AAC32546.1 | |||
| AJ131711.1 | CAB38082.1 | |||
| AK160226.1 | BAE35701.1 | |||
| BC138871.1 | ||||
| BC141142.1 | ||||
| BC145449.1 | ||||
| BC145450.1 | ||||
| BC145451.1 | ||||
| U92883.1 | AAC32540.1 | |||
| U92884.1 | AAC32541.1 |
More Information
Set Membership: |
|

Genomic Map