Assay Details
Target Gene Details
Entrez Gene ID: | 22064 |
Gene Name: | transient receptor potential cation channel, subfamily C, member 2 |
Gene Aliases: |
3010009O07Rik, AI115608, TRPC2a, TRPC2b, Trrp2, mTrp2, smTRPC2, trp2 |
Location: |
Chr.7:102083116-102096864 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 5 - Exon 6 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Trpc2 | NM_001109897.2 | NP_001103367.1 | ||
| AF230803.1 | AAG29951.1 |
Target Gene Details
Entrez Gene ID: | 102443351 |
Gene Name: | Xndc1-transient receptor potential cation channel, subfamily C, member 2 readthrough |
Gene Aliases: |
Xndr-trpc2 |
Location: |
Chr.7:102065491-102096864 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 15 - Exon 16 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Xntrpc | NM_011644.3 | NP_035774.2 | ||
| NR_104582.1 | ||||
| AK040538.1 |
More Information
Set Membership: |
|

Genomic Map