Assay Details
Target Gene Details
Entrez Gene ID: | 102443350 |
Gene Name: | Xrcc1 N-terminal domain containing 1 |
Gene Aliases: |
Xndr |
Location: |
Chr.7:102065491-102083762 on Build GRCm38 |
Assay Gene Location: | Within Exon 8 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Xndc1 | NM_001286689.1 | 8 | 1191 | NP_001273618.1 |
| NM_001286690.1 | 7 | 1096 | NP_001273619.1 | |
| AK041300.1 | 8 | 925 | BAC30894.1 | |
| BC067003.1 | 7 | 1076 | AAH67003.1 |
Target Gene Details
Entrez Gene ID: | 102443351 |
Gene Name: | Xndc1-transient receptor potential cation channel, subfamily C, member 2 readthrough |
Gene Aliases: |
Xndr-trpc2 |
Location: |
Chr.7:102065491-102096864 on Build GRCm38 |
Assay Gene Location: | Within Exon 7 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Xntrpc | NM_011644.3 | 6 | 750 | NP_035774.2 |
| NR_104582.1 | 7 | 1064 | ||
| AK040538.1 | 7 | 842 |

Genomic Map