Assay Details
Target Gene Details
Entrez Gene ID: | 246787 |
Gene Name: | solute carrier family 5 (sodium/glucose cotransporter), member 2 |
Gene Aliases: |
Sglt2 |
Location: |
Chr.7:128265697-128272433 on Build GRCm38 |
Assay Gene Location: | Within Exon 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Slc5a2 | NM_133254.3 | 4 | 374 | NP_573517.1 |
| AK128900.1 | 4 | 388 | ||
| AY033886.1 | 4 | 384 | AAK61606.1 | |
| BC022226.1 | 4 | 374 | AAH22226.1 |
More Information
Set Membership: |
|

Genomic Map