Assay Details
Target Gene Details
Entrez Gene ID: | 20646 |
Gene Name: | small nuclear ribonucleoprotein N |
Gene Aliases: |
2410045I01Rik, HCERN3, Peg4, Pwcr1, SMN |
Location: |
Chr.7:59982502-60140219 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 6 - Exon 6 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Snrpn | NM_001082961.1 | NP_001076430.1 | ||
| NM_001082962.1 | NP_001076431.1 | |||
| NM_013670.3 | NP_038698.1 | |||
| AK010671.1 | ||||
| AK013607.1 | BAB28927.1 | |||
| AK144386.1 | BAE25860.1 | |||
| BC019589.1 | AAH19589.1 | |||
| BC024880.1 | AAH24880.1 | |||
| BQ042870.1 | ||||
| BU614958.1 | ||||
| BY246809.1 | ||||
| BY269658.1 |
Target Gene Details
Entrez Gene ID: | 84704 |
Gene Name: | SNRPN upstream reading frame |
Gene Aliases: |
2410045I01Rik, Snrpn |
Location: |
Chr.7:59982501-60005156 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 2 - Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Snurf | NM_033174.3 | NP_149409.1 | ||
| AF101042.1 | AAD31389.1 | |||
| BC132082.1 | ||||
| BC132084.1 | ||||
| BY271285.1 | ||||
| BY280823.1 |
More Information
Set Membership: |
|

Genomic Map