Assay Details
Target Gene Details
Entrez Gene ID: | 233204 |
Gene Name: | TBC1 domain family, member 17 |
Gene Aliases: |
BC017607 |
Location: |
Chr.7:44840776-44849079 on Build GRCm38 |
Assay Gene Location: | Within Intron 13 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Tbc1d17 | NM_001042655.1 | NP_001036120.1 | ||
| AK039566.1 | BAC30387.1 | |||
| AK046883.1 | BAC32907.1 | |||
| AK081318.1 | ||||
| AK083153.1 | ||||
| AK136705.1 | BAE23103.1 | |||
| AK149917.1 | BAE29165.1 | |||
| AK153732.1 | BAE32159.1 | |||
| AK170821.1 | BAE42052.1 |
More Information
Set Membership: |
|

Genomic Map