Assay Details
Target Gene Details
Entrez Gene ID: | 68472 |
Gene Name: | transmembrane protein 126B |
Gene Aliases: |
1110001A23Rik |
Location: |
Chr.7:90468833-90475995 on Build GRCm38 |
Assay Gene Location: | Within Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Tmem126b | NM_026734.1 | 3 | 274 | NP_081010.1 |
| AK003201.1 | 3 | 275 | BAB22637.1 | |
| BC049680.1 | 3 | 274 | AAH49680.1 |

Genomic Map